missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzyl nicotinate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1850518
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Benzyl nicotinate | |
21°C to 23°C | |
179°C to 180°C (5 mmHg) | |
C13H11NO2 | |
MFCD00023584 | |
159169 | |
benzyl nicotinate, nicotinic acid benzyl ester, rubriment, pycaril, pykaryl, niacin benzyl ester, 3-pyridinecarboxylic acid, phenylmethyl ester, benzylis nicotinas, nicotinic acid, benzyl ester, nicotinsaeurebenzylester | |
C1=CC=C(C=C1)COC(=O)C2=CN=CC=C2 | |
213.236 | |
CHEBI:31268 | |
99% |
94-44-0 | |
1.165 | |
170°C (338°F) | |
1.57 | |
50 g | |
14,6526 | |
KVYGGMBOZFWZBQ-UHFFFAOYSA-N | |
benzyl pyridine-3-carboxylate | |
7191 | |
213.24 |
Chemical Identifiers
94-44-0 | |
213.236 | |
KVYGGMBOZFWZBQ-UHFFFAOYSA-N | |
7191 | |
benzyl pyridine-3-carboxylate |
C13H11NO2 | |
MFCD00023584 | |
benzyl nicotinate, nicotinic acid benzyl ester, rubriment, pycaril, pykaryl, niacin benzyl ester, 3-pyridinecarboxylic acid, phenylmethyl ester, benzylis nicotinas, nicotinic acid, benzyl ester, nicotinsaeurebenzylester | |
CHEBI:31268 | |
C1=CC=C(C=C1)COC(=O)C2=CN=CC=C2 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 202-332-3
RTECSNumber : QT0850000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only