missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzyl nicotinate, 98%, Thermo Scientific Chemicals

$102.08 - $371.59

Chemical Identifiers

CAS 94-44-0
Molecular Formula C13H11NO2
Molecular Weight (g/mol) 213.236
MDL Number MFCD00023584
InChI Key KVYGGMBOZFWZBQ-UHFFFAOYSA-N
Synonym benzyl nicotinate, nicotinic acid benzyl ester, rubriment, pycaril, pykaryl, niacin benzyl ester, 3-pyridinecarboxylic acid, phenylmethyl ester, benzylis nicotinas, nicotinic acid, benzyl ester, nicotinsaeurebenzylester
PubChem CID 7191
ChEBI CHEBI:31268
IUPAC Name benzyl pyridine-3-carboxylate
SMILES C1=CC=C(C=C1)COC(=O)C2=CN=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1850518
View Documents
Thermo Scientific Chemicals
A1850518
50 g
Each for $102.08
 
AAA1850530
View Documents
Thermo Scientific Chemicals
A1850530
250 g
Each for $371.59
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

94-44-0
213.236
KVYGGMBOZFWZBQ-UHFFFAOYSA-N
7191
benzyl pyridine-3-carboxylate
C13H11NO2
MFCD00023584
benzyl nicotinate, nicotinic acid benzyl ester, rubriment, pycaril, pykaryl, niacin benzyl ester, 3-pyridinecarboxylic acid, phenylmethyl ester, benzylis nicotinas, nicotinic acid, benzyl ester, nicotinsaeurebenzylester
CHEBI:31268
C1=CC=C(C=C1)COC(=O)C2=CN=CC=C2

Specifications

94-44-0
1.165
170°C (338°F)
1.57
50 g
14,6526
KVYGGMBOZFWZBQ-UHFFFAOYSA-N
benzyl pyridine-3-carboxylate
7191
213.24
Benzyl nicotinate
21°C to 23°C
179°C to 180°C (5 mmHg)
C13H11NO2
MFCD00023584
159169
benzyl nicotinate, nicotinic acid benzyl ester, rubriment, pycaril, pykaryl, niacin benzyl ester, 3-pyridinecarboxylic acid, phenylmethyl ester, benzylis nicotinas, nicotinic acid, benzyl ester, nicotinsaeurebenzylester
C1=CC=C(C=C1)COC(=O)C2=CN=CC=C2
213.236
CHEBI:31268
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

EINECSNumber : 202-332-3

RTECSNumber : QT0850000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only