Bio-Serv™ Mouse Igloo™ Rodent Enrichment Device, Certified DFS Item
Provides shelter and a secluded place to build a nest
Supplier:  Bio-Serv™ K3327
 View more versions of this product
Catalog No. 36-099-3893
Explore available promotions
Description
Description
Provides shelter and a secluded place to build a nest
  • Transparent material provides ease of visibility
  • Great addition to breeding programs
  • Fits most shoebox cages
  • Stacks easily for convenient storage
  • Made of high temperature polycarbonate
  • Autoclavable
  • Cagewasher safe
  • Certified (Contaminant screened)
Specifications
Specifications
Mouse Igloo
Red
Provide shelter and a secluded place to build a nest, great addition to breeding programs
5.7 cm
Autoclavable
Polycarbonate
2.25 in.
Product Suggestions
Product Suggestions