bioMérieux™ VITEK™ Test Cards DFS Item
Species-level identification and resistance detection in bacteria and yeast
Supplier:  bioMérieux™ 21342
 View more versions of this product
Catalog No. B21342
Explore available promotions
Description
Description
  • Inoculation with a simple, standardized saline suspension of organism
  • No reagent addition
  • Closed and disposable for optimal user safety
  • Pre-applied bar codes for maximum traceability
  • Highest level of automation
Specifications
Specifications
Identification Test
Gram positive cocci
Vitek™ 2 colorimetric
20 Pack