missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

bioMérieux™ VITEK™ Test Cards DFS Item

Species-level identification and resistance detection in bacteria and yeast

$725.34 - $725.34

Specifications

Product Type Identification Test
For Use With (Equipment) Vitek™ 2 colorimetric
Quantity 20 Pack
View More Specs

Products 2
Catalog Number Mfr. No. For Use With (Application) Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Price Quantity  
B21343 DFS Item
View Documents
bioMérieux™
21343
Yeast
Pack of 20 Each for $725.34
 
B21342 DFS Item
View Documents
bioMérieux™
21342
Gram positive cocci
Pack of 20 Each for $725.34
 
Description

Description

  • Inoculation with a simple, standardized saline suspension of organism
  • No reagent addition
  • Closed and disposable for optimal user safety
  • Pre-applied bar codes for maximum traceability
  • Highest level of automation

Specifications

Specifications

Identification Test
Vitek™ 2 colorimetric
20 Pack
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'