missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ BIOShield™ 720 Rotor Tube Adapters

Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes.

Supplier:  Thermo Scientific™ 75003701

 View more versions of this product

Catalog No. 75-003-701


Description

Description

Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes. These adapters are for use with the BIOShield 720 High Speed Swinging-Bucket Rotor.

  • Maximum tube capacity: 4 × 150mL
  • Maximum O.D. × L: 50 × 122mm
Specifications

Specifications

Rotor Tube Adapter
10 mL
10 mL
Blood Collection
10mL Blood Collection
SDS
Documents

Documents

Promotions

Promotions