Thermo Scientific™ BIOShield™ 720 Rotor Tube Adapters
Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes.
$793.75 - $924.04
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity | |||||
75-003-677
|
Thermo Scientific™
75003677 |
50mL Conical |
Each for $827.04
|
|
|||||
75-003-678
|
Thermo Scientific™
75003678 |
15mL Conical |
Each for $924.04
|
|
|||||
75-003-693
|
Thermo Scientific™
75003693 |
180mL Polypropylene Screw Cap (Bottle) |
Each for $875.17
|
|
|||||
75-003-701
|
Thermo Scientific™
75003701 |
10mL Blood Collection |
Each for $877.02
|
|
|||||
75-003-813
|
Thermo Scientific™
75003813 |
150mL Round-bottom, Open-tube |
Each for $875.17
|
|
|||||
75-003-814
|
Thermo Scientific™
75003814 |
100mL Round-bottom, Open-tube |
Each for $793.75
|
|
|||||
75-003-816
|
Thermo Scientific™
75003816 |
50mL DIN Round-bottom |
Each for $827.04
|
|
|||||
75-003-817
|
Thermo Scientific™
75003817 |
25mL DIN Round-bottom |
Each for $793.75
|
|
|||||
75-003-818
|
Thermo Scientific™
75003818 |
25mL Universal |
Each for $914.02
|
|
|||||
75-003-820
|
Thermo Scientific™
75003820 |
15mL Starstedt |
Each for $827.04
|
|
|||||
75-003-821
|
Thermo Scientific™
75003821 |
5/7mL Blood Collection |
Each for $889.60
|
|
|||||
75-003-822
|
Thermo Scientific™
75003822 |
5/7mL Round-bottom, Open-tube |
Each for $867.63
|
|
|||||
75-003-823
|
Thermo Scientific™
75003823 |
1.5/2.0mL Conical Microtube |
Each for $793.75
|
|
|||||
Description
Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes. These adapters are for use with the BIOShield 720 High Speed Swinging-Bucket Rotor.
- Maximum tube capacity: 4 × 150mL
- Maximum O.D. × L: 50 × 122mm