Thermo Scientific™ BIOShield™ 720 Rotor Tube Adapters
Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes.
Supplier:  Thermo Scientific™ 75003817
 View more versions of this product
Catalog No. 75-003-817
Description
Description
Enhance your rotor's versatility with Thermo Scientific™ BIOShield™ 720 Tube Adapters, which support a wide assortment of tube sizes. These adapters are for use with the BIOShield 720 High Speed Swinging-Bucket Rotor.
  • Maximum tube capacity: 4 × 150mL
  • Maximum O.D. × L: 50 × 122mm
Specifications
Specifications
25mL DIN Round-bottom