missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Biphenyl-2-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3232006

 View more versions of this product

Catalog No. AAH3232006


Explore available promotions

Description

Description

Biphenyl-2-boronic acid pinacol ester, is used as a main benefits of the use of boronic esters in Suzuki-Miyaura cross-couplings- Enabling the coupling of organoboronic acids that are sensitive to hydrolytic deboronation. It includes other additional application such as a number of chiral boronic esters, such as the ones illustrated hereafter, have been used as inducers in stereoselective reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Biphenyl-2-boronic acid pinacol ester, is used as a main benefits of the use of boronic esters in Suzuki-Miyaura cross-couplings- Enabling the coupling of organoboronic acids that are sensitive to hydrolytic deboronation. It includes other additional application such as a number of chiral boronic esters, such as the ones illustrated hereafter, have been used as inducers in stereoselective reactions.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Specifications

Specifications

Biphenyl-2-boronic acid pinacol ester
76°C to 79°C
MFCD14636459
biphenyl-2-boronic acid pinacol ester, 2-1,1'-biphenyl-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-biphenylyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
WCXWQEUBHZKNMQ-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-(2-phenylphenyl)-1,3,2-dioxaborolane
59207619
97%
914675-52-8
C18H21BO2
5 g
Insoluble in water. Soluble in organic solvents.
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C3=CC=CC=C3
280.174
280.17

Chemical Identifiers

914675-52-8
280.174
WCXWQEUBHZKNMQ-UHFFFAOYSA-N
59207619
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2C3=CC=CC=C3
C18H21BO2
MFCD14636459
biphenyl-2-boronic acid pinacol ester, 2-1,1'-biphenyl-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-2-biphenylyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
4,4,5,5-tetramethyl-2-(2-phenylphenyl)-1,3,2-dioxaborolane
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only