missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(1,5-cyclooctadiene)nickel(0), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 223970050

 View more versions of this product

Catalog No. AC223970050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Bis(1, 5-cyclooctadiene)nickel(0)
60.0°C
Glass bottle
MFCD00058902
bis 1,5-cyclooctadiene nickel 0, bis 1,5-cyclooctadiene nickel, ni cod 2, ccris 5973, bis cyclooctadiene nickel 0, nickel, bis 1,5-cyclooctadiene, bis cyclooctadiene nickel, 1z,5z-cycloocta-1,5-diene; nickel, nickel biscod, pubchem16018
JRTIUDXYIUKIIE-KZUMESAESA-N
(1Z,5Z)-cycloocta-1,5-diene;nickel
6433264
Shiny Crystalline Powder
1295-35-8
Golden
C16H24Ni
5 g
Solubility in water: insoluble.
[Ni].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
275.06
275.06

Chemical Identifiers

1295-35-8
275.06
JRTIUDXYIUKIIE-KZUMESAESA-N
6433264
C16H24Ni
MFCD00058902
bis 1,5-cyclooctadiene nickel 0, bis 1,5-cyclooctadiene nickel, ni cod 2, ccris 5973, bis cyclooctadiene nickel 0, nickel, bis 1,5-cyclooctadiene, bis cyclooctadiene nickel, 1z,5z-cycloocta-1,5-diene; nickel, nickel biscod, pubchem16018
[Ni].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.
May cause cancer.
May cause an allergic skin reaction.

GHS P Statement
Do not handle until all safety precautions have been read and understood.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Use personal protective equipment as required.
IF exposed or concerned:

GHS Signal Word: Danger

EINECSNumber : 215-072-

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only