Learn More
Bis(1,5-cyclooctadiene)nickel(0), Thermo Scientific Chemicals
$145.91 - $487.49
Chemical Identifiers
CAS | 1295-35-8 |
---|---|
Molecular Formula | C16H24Ni |
Molecular Weight (g/mol) | 275.06 |
MDL Number | MFCD00058902 |
InChI Key | JRTIUDXYIUKIIE-KZUMESAESA-N |
Synonym | bis 1,5-cyclooctadiene nickel 0, bis 1,5-cyclooctadiene nickel, ni cod 2, ccris 5973, bis cyclooctadiene nickel 0, nickel, bis 1,5-cyclooctadiene, bis cyclooctadiene nickel, 1z,5z-cycloocta-1,5-diene; nickel, nickel biscod, pubchem16018 |
PubChem CID | 6433264 |
SMILES | [Ni].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC223970010
|
Thermo Scientific Chemicals
223970010 |
1 g |
Each for $145.91
|
|
|||||
AC223970050
|
Thermo Scientific Chemicals
223970050 |
5 g |
Each for $487.49
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1295-35-8 | |
275.06 | |
JRTIUDXYIUKIIE-KZUMESAESA-N | |
6433264 |
C16H24Ni | |
MFCD00058902 | |
bis 1,5-cyclooctadiene nickel 0, bis 1,5-cyclooctadiene nickel, ni cod 2, ccris 5973, bis cyclooctadiene nickel 0, nickel, bis 1,5-cyclooctadiene, bis cyclooctadiene nickel, 1z,5z-cycloocta-1,5-diene; nickel, nickel biscod, pubchem16018 | |
[Ni].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1 |
Specifications
1295-35-8 | |
Golden | |
C16H24Ni | |
1 g | |
Solubility in water: insoluble. | |
[Ni].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1 | |
275.06 | |
275.06 | |
Bis(1, 5-cyclooctadiene)nickel(0) |
60.0°C | |
Glass bottle | |
MFCD00058902 | |
bis 1,5-cyclooctadiene nickel 0, bis 1,5-cyclooctadiene nickel, ni cod 2, ccris 5973, bis cyclooctadiene nickel 0, nickel, bis 1,5-cyclooctadiene, bis cyclooctadiene nickel, 1z,5z-cycloocta-1,5-diene; nickel, nickel biscod, pubchem16018 | |
JRTIUDXYIUKIIE-KZUMESAESA-N | |
(1Z,5Z)-cycloocta-1,5-diene;nickel | |
6433264 | |
Shiny Crystalline Powder |
Safety and Handling
GHS H Statement
Flammable solid.
May cause cancer.
May cause an allergic skin reaction.
GHS P Statement
Do not handle until all safety precautions have been read and understood.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Use personal protective equipment as required.
IF exposed or concerned:
GHS Signal Word: Danger
EINECSNumber : 215-072-
RUO – Research Use Only