missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II), 98%, Thermo Scientific Chemicals

NMR shift reagent, nickel oxide precursor

Supplier:  Thermo Scientific Chemicals 4047006

 View more versions of this product

Catalog No. AA4047006



Description

Description

NMR shift reagent, nickel oxide precursor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II)
223°C to 225°C
C22H38NiO4
5 g
FPQQRRXEOLXSCJ-UHFFFAOYSA-N
(E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;nickel
131675868
98%
14481-08-4
90°C to 110°C (0.1 mmHg)(sublimation)
MFCD00192348
bis 2,2,6,6-tetramethyl-3,5-heptanedionate nickel ii, bis 2,2,6,6-tetramethyl-3,5-heptanedionato nickel ii , min-ni ni tmhd 2
[Ni++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C
425.24
425.24
Powder

Chemical Identifiers

14481-08-4
425.24
FPQQRRXEOLXSCJ-UHFFFAOYSA-N
131675868
[Ni++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C
C22H38NiO4
MFCD00192348
bis 2,2,6,6-tetramethyl-3,5-heptanedionate nickel ii, bis 2,2,6,6-tetramethyl-3,5-heptanedionato nickel ii , min-ni ni tmhd 2
(E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;nickel
Safety and Handling

Safety and Handling

GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H350

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only