missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II), 98%, Thermo Scientific Chemicals
NMR shift reagent, nickel oxide precursor
$209.10 - $916.26
Chemical Identifiers
CAS | 14481-08-4 |
---|---|
Molecular Formula | C22H38NiO4 |
Molecular Weight (g/mol) | 425.24 |
MDL Number | MFCD00192348 |
InChI Key | FPQQRRXEOLXSCJ-UHFFFAOYSA-N |
Synonym | bis 2,2,6,6-tetramethyl-3,5-heptanedionate nickel ii, bis 2,2,6,6-tetramethyl-3,5-heptanedionato nickel ii , min-ni ni tmhd 2 |
PubChem CID | 131675868 |
IUPAC Name | (E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;nickel |
SMILES | [Ni++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C |
Description
NMR shift reagent, nickel oxide precursor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14481-08-4 | |
425.24 | |
FPQQRRXEOLXSCJ-UHFFFAOYSA-N | |
131675868 | |
[Ni++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C |
C22H38NiO4 | |
MFCD00192348 | |
bis 2,2,6,6-tetramethyl-3,5-heptanedionate nickel ii, bis 2,2,6,6-tetramethyl-3,5-heptanedionato nickel ii , min-ni ni tmhd 2 | |
(E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;nickel |
Specifications
14481-08-4 | |
90°C to 110°C (0.1 mmHg)(sublimation) | |
MFCD00192348 | |
bis 2,2,6,6-tetramethyl-3,5-heptanedionate nickel ii, bis 2,2,6,6-tetramethyl-3,5-heptanedionato nickel ii , min-ni ni tmhd 2 | |
[Ni++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C | |
425.24 | |
425.24 | |
Powder |
223°C to 225°C | |
C22H38NiO4 | |
1 g | |
FPQQRRXEOLXSCJ-UHFFFAOYSA-N | |
(E)-5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;nickel | |
131675868 | |
98% | |
Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)nickel(II) |
Safety and Handling
GHS H Statement
H350-H317
May cause cancer.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only