missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(4-morpholinyl)methane, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5113306

 View more versions of this product

Catalog No. AAH5113306


Explore available promotions

Description

Description

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

Bis(4-morpholinyl)methane
122°C to 124°C (12 mmHg)
MFCD00023369
dimorpholinomethane, n,n'-dimorpholinomethane, 4,4-methylenedimorpholine, 4,4'-methylenedimorpholine, bismorpholino methane, n,n'-methylenebismorpholine, morpholine, 4,4'-methylenebis, unii-7o79dzw79z, bis morpholino-methan german, bis 4-morpholinyl methane
MIFZZKZNMWTHJK-UHFFFAOYSA-N
4-(morpholin-4-ylmethyl)morpholine
21839
98%
5625-90-1
C9H18N2O2
5 g
Soluble in water.
C1COCCN1CN2CCOCC2
186.255
186.25

Chemical Identifiers

5625-90-1
186.255
MIFZZKZNMWTHJK-UHFFFAOYSA-N
21839
C1COCCN1CN2CCOCC2
C9H18N2O2
MFCD00023369
dimorpholinomethane, n,n'-dimorpholinomethane, 4,4-methylenedimorpholine, 4,4'-methylenedimorpholine, bismorpholino methane, n,n'-methylenebismorpholine, morpholine, 4,4'-methylenebis, unii-7o79dzw79z, bis morpholino-methan german, bis 4-morpholinyl methane
4-(morpholin-4-ylmethyl)morpholine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H315
Causes serious eye damage.
Causes skin irritation.

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H302+H312+H332-H314-H317-H341-H350-H373

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only