missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(4-morpholinyl)methane, 98%, Thermo Scientific Chemicals

$55.48 - $193.84

Chemical Identifiers

CAS 5625-90-1
Molecular Formula C9H18N2O2
Molecular Weight (g/mol) 186.255
MDL Number MFCD00023369
InChI Key MIFZZKZNMWTHJK-UHFFFAOYSA-N
Synonym dimorpholinomethane, n,n'-dimorpholinomethane, 4,4-methylenedimorpholine, 4,4'-methylenedimorpholine, bismorpholino methane, n,n'-methylenebismorpholine, morpholine, 4,4'-methylenebis, unii-7o79dzw79z, bis morpholino-methan german, bis 4-morpholinyl methane
PubChem CID 21839
IUPAC Name 4-(morpholin-4-ylmethyl)morpholine
SMILES C1COCCN1CN2CCOCC2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5113306
View Documents
Thermo Scientific Chemicals
H5113306
5 g
Each for $55.48
 
AAH5113314
View Documents
Thermo Scientific Chemicals
H5113314
25 g
Each for $193.84
 
Description

Description

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Chemical Identifiers

5625-90-1
186.255
MIFZZKZNMWTHJK-UHFFFAOYSA-N
21839
C1COCCN1CN2CCOCC2
C9H18N2O2
MFCD00023369
dimorpholinomethane, n,n'-dimorpholinomethane, 4,4-methylenedimorpholine, 4,4'-methylenedimorpholine, bismorpholino methane, n,n'-methylenebismorpholine, morpholine, 4,4'-methylenebis, unii-7o79dzw79z, bis morpholino-methan german, bis 4-morpholinyl methane
4-(morpholin-4-ylmethyl)morpholine

Specifications

5625-90-1
C9H18N2O2
5 g
Soluble in water.
C1COCCN1CN2CCOCC2
186.255
186.25
Bis(4-morpholinyl)methane
122°C to 124°C (12 mmHg)
MFCD00023369
dimorpholinomethane, n,n'-dimorpholinomethane, 4,4-methylenedimorpholine, 4,4'-methylenedimorpholine, bismorpholino methane, n,n'-methylenebismorpholine, morpholine, 4,4'-methylenebis, unii-7o79dzw79z, bis morpholino-methan german, bis 4-morpholinyl methane
MIFZZKZNMWTHJK-UHFFFAOYSA-N
4-(morpholin-4-ylmethyl)morpholine
21839
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H315
Causes serious eye damage.
Causes skin irritation.

P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P333+P313-P363-P501c

H302+H312+H332-H314-H317-H341-H350-H373

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only