missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(methylthio)methane 98.0+%, TCI America™

Supplier:  TCI America B120425ML

 View more versions of this product

Catalog No. B120425ML


Explore available promotions

Description

Description

Specifications

Specifications

Bis(methylthio)methane
152°C
MFCD00008564
1993
LOCDPORVFVOGCR-UHFFFAOYSA-N
bis(methylsulfanyl)methane
15380
≥98.0% (GC)
1618-26-4
C3H8S2
25 mL
bis methylthio methane, bis methylsulfanyl methane, 2,4-dithiapentane, methane, bis methylthio, bis methylmercapto methane, formaldehyde dimethyl mercaptal, methylenebis methyl sulfide, thioformaldehyde dimethylacetal, ch3sch2sch3
CSCSC
108.217
108.22
Liquid

Chemical Identifiers

1618-26-4
108.217
LOCDPORVFVOGCR-UHFFFAOYSA-N
15380
CSCSC
C3H8S2
MFCD00008564
bis methylthio methane, bis methylsulfanyl methane, 2,4-dithiapentane, methane, bis methylthio, bis methylmercapto methane, formaldehyde dimethyl mercaptal, methylenebis methyl sulfide, thioformaldehyde dimethylacetal, ch3sch2sch3
bis(methylsulfanyl)methane
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions