missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bis(methylthio)methane 98.0+%, TCI America™

$43.25 - $147.06

Chemical Identifiers

CAS 1618-26-4
Molecular Formula C3H8S2
Molecular Weight (g/mol) 108.217
MDL Number MFCD00008564
InChI Key LOCDPORVFVOGCR-UHFFFAOYSA-N
Synonym bis methylthio methane, bis methylsulfanyl methane, 2,4-dithiapentane, methane, bis methylthio, bis methylmercapto methane, formaldehyde dimethyl mercaptal, methylenebis methyl sulfide, thioformaldehyde dimethylacetal, ch3sch2sch3
PubChem CID 15380
IUPAC Name bis(methylsulfanyl)methane
SMILES CSCSC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B12045ML
View Documents
TCI America
B12045ML
5 mL
Each for $43.25
 
B120425ML
View Documents
TCI America
B120425ML
25 mL
Each for $147.06
 
Description

Description

Specifications

Chemical Identifiers

1618-26-4
108.217
LOCDPORVFVOGCR-UHFFFAOYSA-N
15380
CSCSC
C3H8S2
MFCD00008564
bis methylthio methane, bis methylsulfanyl methane, 2,4-dithiapentane, methane, bis methylthio, bis methylmercapto methane, formaldehyde dimethyl mercaptal, methylenebis methyl sulfide, thioformaldehyde dimethylacetal, ch3sch2sch3
bis(methylsulfanyl)methane

Specifications

1618-26-4
C3H8S2
5 mL
bis methylthio methane, bis methylsulfanyl methane, 2,4-dithiapentane, methane, bis methylthio, bis methylmercapto methane, formaldehyde dimethyl mercaptal, methylenebis methyl sulfide, thioformaldehyde dimethylacetal, ch3sch2sch3
CSCSC
108.217
108.22
Liquid
152°C
MFCD00008564
1993
LOCDPORVFVOGCR-UHFFFAOYSA-N
bis(methylsulfanyl)methane
15380
≥98.0% (GC)
Bis(methylthio)methane
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'