missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bismuth hydroxide, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 393290500

 View more versions of this product

Catalog No. AC393290500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Bismuth hydroxide
White to Cream
Powder
BiH3O3
MFCD00046188
Solubility in water: negligible. Other solubilities: soluble in acids
[OH-].[OH-].[OH-].[Bi+3]
260.00
Positive
Glass bottle
415.0°C
10361-43-0
50 g
Bi(OH)3
15, 1267
TZSXPYWRDWEXHG-UHFFFAOYSA-K
bismuth(3+) trihydroxide
260
1.0% max.

Chemical Identifiers

10361-43-0
260.00
TZSXPYWRDWEXHG-UHFFFAOYSA-K
[OH-].[OH-].[OH-].[Bi+3]
BiH3O3
MFCD00046188
bismuth(3+) trihydroxide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 233-790-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only