missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bismuth hydroxide, Thermo Scientific Chemicals
$70.89 - $70.89
Chemical Identifiers
CAS | 10361-43-0 |
---|---|
Molecular Formula | BiH3O3 |
Molecular Weight (g/mol) | 260.00 |
MDL Number | MFCD00046188 |
InChI Key | TZSXPYWRDWEXHG-UHFFFAOYSA-K |
IUPAC Name | bismuth(3+) trihydroxide |
SMILES | [OH-].[OH-].[OH-].[Bi+3] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC393290500
|
Thermo Scientific Chemicals
393290500 |
50 g | Glass bottle |
Each for $70.89
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10361-43-0 | |
260.00 | |
TZSXPYWRDWEXHG-UHFFFAOYSA-K | |
[OH-].[OH-].[OH-].[Bi+3] |
BiH3O3 | |
MFCD00046188 | |
bismuth(3+) trihydroxide |
Specifications
415.0°C | |
10361-43-0 | |
50 g | |
Bi(OH)3 | |
15, 1267 | |
TZSXPYWRDWEXHG-UHFFFAOYSA-K | |
bismuth(3+) trihydroxide | |
260 | |
1.0% max. | |
Bismuth hydroxide |
White to Cream | |
Powder | |
BiH3O3 | |
MFCD00046188 | |
Solubility in water: negligible. Other solubilities: soluble in acids | |
[OH-].[OH-].[OH-].[Bi+3] | |
260.00 | |
Positive | |
Glass bottle |
Safety and Handling
EINECSNumber : 233-790-2
RUO – Research Use Only