missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) iodide, Puratronic™, 99.999% (metals basis)

CAS: 7787-64-6 | BiI3 | 589.69 g/mol

$85.45 - $1011.52

Chemical Identifiers

CAS 7787-64-6
Molecular Formula BiI3
Molecular Weight (g/mol) 589.69
MDL Number MFCD00010894
InChI Key KOECRLKKXSXCPB-UHFFFAOYSA-K
Synonym bismuth iodide, bismuth iii iodide, bismuth triiodide, bismuthine, triiodo, triiodobismuthine, triiodobismuth, bismuth iodide,, ksc179g5d
PubChem CID 111042
SMILES [I-].[I-].[I-].[Bi+3]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1065604
View Documents
Thermo Scientific Chemicals
01065604
2 g
Each for $85.45
Only null left
 
AA1065609
View Documents
Thermo Scientific Chemicals
01065609
10 g
Each for $287.74
Only null left
 
AA1065618
View Documents
Thermo Scientific Chemicals
01065618
50 g
Each for $1,011.52
Only null left
 
Description

Description

Polycrystalline films of bismuth triiodide are used as beta direct and digital detectors for medical imaging applications. Bismuth(III) iodide is used as an ice nucleant. It is also used in electron microscopy to reveal synapses. Addition of bismuth triiodide has been shown to improve the glass forming ability and thermal stability of Ge-Te-BiI(3) chalcogenide glasses. Solution-processable BiI3 nanosheets have been shown to exhibit the potential to be used as HTLs (Hole Transport Layer) in conventional OPV (organic photovoltaic) devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7787-64-6
589.69
KOECRLKKXSXCPB-UHFFFAOYSA-K
111042
BiI3
MFCD00010894
bismuth iodide, bismuth iii iodide, bismuth triiodide, bismuthine, triiodo, triiodobismuthine, triiodobismuth, bismuth iodide,, ksc179g5d
[I-].[I-].[I-].[Bi+3]

Specifications

∼500°C
7787-64-6
Lump/Powder
BiI3
UN3260
bismuth iodide, bismuth iii iodide, bismuth triiodide, bismuthine, triiodo, triiodobismuthine, triiodobismuth, bismuth iodide,, ksc179g5d
KOECRLKKXSXCPB-UHFFFAOYSA-K
triiodobismuthane
111042
99.999%
Acidic
5.778 g/mL
(metals basis)
408°C
Gray to Black
2 g
MFCD00010894
14,1269
Insoluble in water. Highly soluble in ethanol,benzene,toluene and xylene,hydrochloric acid and in hydriodic acid.
[I-].[I-].[I-].[Bi+3]
589.69
589.69
Puratronic
Hygroscopic; Light sensitive
Bismuth(III) iodide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 232-127-4

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only