Fisherbrand™ Black Phenolic Caps
Black phenolic screw cap with poly-seal liner is manufactured from low density polyethylene. For use with various Fisherbrand molded glass bottles
Description
Unique cone liner design offers superior torque retention and excellent sealing characteristicsSpecifications
Black | |
Black | |
No | |
Polyethylene Cone |
576 | |
15-425 | |
Lined Cap | |
Phenolic |