Fisherbrand™ Black Phenolic Caps
Black phenolic screw cap with poly-seal liner is manufactured from low density polyethylene. For use with various Fisherbrand molded glass bottles
$13.43 - $2592.52
Specifications
Closure Color | Black |
---|---|
Closure Liner | Polyethylene Cone |
Closure Material | Phenolic |
Catalog Number | Mfr. No. | Neck Finish | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Neck Finish | Quantity | Price | Quantity | ||||
FB02911727A
|
N/A | 38-400 | 1600 |
Case of 1600 Each for $2,227.43
|
|
||||
FB02912054
|
N/A | 20-400 | 576 |
Case of 576 Each for $669.64
|
|
||||
FB02911803
|
N/A | 20-400 | 5500 |
Case of 5500 Each for $2,473.04
|
|
||||
FB02912055
|
N/A | 24-400 | 12 |
Pack of 12 Each for $13.43
Case of 48 Pack for $644.64
|
|
||||
FB02912056
|
N/A | 22-400 | 12 |
Pack of 12 Each for $16.11
Case of 48 Pack for $773.28
|
|
||||
FB02912059
|
N/A | 18-400 | 12 |
Pack of 12 Each for $13.79
Case of 48 Pack for $661.92
|
|
||||
FB02912058
|
N/A | 15-425 | 576 |
Case of 576 Each for $520.00
|
|
||||
FB02912061
|
N/A | 28-400 | 576 |
Pack of 12 Each for $13.68
Case of 48 Pack for $656.64
|
|
||||
FB02912062
|
N/A | 33-400 | 12 |
Pack of 12 Each for $19.57
Case of 48 Pack for $939.36
|
|
||||
FB02912101
|
N/A | 28-400 | 3100 |
Case of 3100 Each for $2,592.52
|
|
||||
FB02912063A
|
N/A | 38-400 | 12 |
Pack of 12 Each for $19.61
Case of 48 Pack for $941.28
|
|
||||
Description
Unique cone liner design offers superior torque retention and excellent sealing characteristicsSpecifications
Black | |
Polyethylene Cone | |
Phenolic |