Qorpak™ Black Phenolic PolyCone Lined Caps
Forms an exceptionally tight seal and offer a good chemical barrier. Qorpak™ Black Phenolic PolyCone Lined Caps is used for everything from liquids, to soaps, to macro-invertebrates.
Supplier: Qorpak™ CAP00204
Description
- Molded liner around the sealing areas eliminates leakage, evaporation, contamination, binding and backing off
- Available in ribbed style
- Available in bulk/bagged packaging
Specifications
Black | |
2200 Cs. | |
33-400 | |
Polycone |
Phenolic Resin | |
Bulk | |
Wide mouth bottle | |
Ribbed |