Qorpak™ Black Phenolic PolyCone Lined Caps
Forms an exceptionally tight seal and offer a good chemical barrier. Qorpak™ Black Phenolic PolyCone Lined Caps is used for everything from liquids, to soaps, to macro-invertebrates.
$806.12 - $3793.76
Specifications
Color | Black |
---|---|
Material | Phenolic Resin |
Closure Liner | Polycone |
Closure Type | Ribbed |
Catalog Number | Mfr. No. | Neck Finish | For Use With (Equipment) | No. per Case | Packaging | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Neck Finish | For Use With (Equipment) | No. per Case | Packaging | Price | Quantity | ||
02-991-076
|
Qorpak™
CAP00154 |
22-400 | Wide mouth bottle | 5000 Cs. | Bulk |
Case of 5000 Each for $3,793.76
|
|
||
36-102-7005
|
Qorpak™
CAP00176 |
24-400 | Wide mouth bottle | 576 Cs. | Bagged |
Case of 576 Each for $819.69
|
|
||
36-101-2194
|
Qorpak™
CAP00212 |
33-400 | Wide mouth bottle | 576 Cs. | Bagged |
Case of 576 Each for $1,124.41
|
|
||
02-991-174
|
Qorpak™
CAP00190 |
28-400 | Wide mouth bottle | 3100 Cs. | Bulk |
Case of 3100 Each for $3,265.97
|
|
||
02-991-097
|
Qorpak™
CAP00204 |
33-400 | Wide mouth bottle | 2200 Cs. | Bulk |
Case of 2200 Each for $2,646.34
|
|
||
02-883-5J
|
Qorpak™
CAP00195 |
28-400 | Wide mouth bottle | 576 Cs. | Bagged |
Case of 576 Each for $806.12
|
|
||
Description
- Molded liner around the sealing areas eliminates leakage, evaporation, contamination, binding and backing off
- Available in ribbed style
- Available in bulk/bagged packaging
Specifications
Black | |
Phenolic Resin | |
Polycone | |
Ribbed |