missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Borane-tert-butylamine complex, 95%, powder, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 177301000

 View more versions of this product

Catalog No. AC177301000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Specifications

Borane-tert-butylamine complex
7337-45-3,75-64-9
White
95%
C4H14BN
MFCD00075635
11,87
Solubility in water: 2.7g/100mL (20°C)
B.CC(C)(C)N
86.97
86.97
Fine Crystalline Powder
95%
96°C to 101°C
Authentic
Glass bottle
(CH3)3CNH2·BH3
100 g
tert-butylamine borane, borane-tert-butylamine complex, borane tert-butylamine complex, borane-tert-butylaminecomplex, c4h14bn, borane t-butylamine, t-butylamine.borane, tert-butylamineborane, borane tert-butylamine, tert-butylamine-borane
GKFJEDWZQZKYHV-UHFFFAOYSA-N
boron;2-methylpropan-2-amine
6364547
95%

Chemical Identifiers

7337-45-3
86.97
GKFJEDWZQZKYHV-UHFFFAOYSA-N
6364547
B.CC(C)(C)N
C4H14BN
MFCD00075635
tert-butylamine borane, borane-tert-butylamine complex, borane tert-butylamine complex, borane-tert-butylaminecomplex, c4h14bn, borane t-butylamine, t-butylamine.borane, tert-butylamineborane, borane tert-butylamine, tert-butylamine-borane
boron;2-methylpropan-2-amine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Toxic in contact with skin.
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Flammable solid.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume

GHS Signal Word: Danger

EINECSNumber : 230-851-5

RTECSNumber : EO3900000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only