missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Borane-tetrahydrofuran complex, 1M solution in THF, Stabilized, AcroSeal™, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 175088000

 View more versions of this product

Catalog No. AC175088000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
Specifications

Specifications

Borane-tetrahydrofuran complex
1M solution in tetrahydrofuran, stabilized
C4H11BO
0.96 to 1.08M
0.8760g/mL
borane-tetrahydrofuran complex, tetrahydrofuran borane, bh3.thf, borane tetrahydrofuran complex solution, borane-d3-thf complex solution, borane-tetrahydrofuran, unii-5ear4err1l, oxolane borane, boron; oxolane, borane thf
RMCYTHFAWCWRFA-UHFFFAOYSA-N
oxolane borane
11062302
AcroSeal™ Glass bottle
Stabilized
−22°C
Colorless
14044-65-6,109-99-9
MFCD00012429
01,199; 02,106; 03,76; 04,124; 05,184; 06,161; 07,89; 12,65; 17,101
15, 1336
Solubility in water: reacts.
B.C1CCOC1
85.94
85.94
0.876
800 mL
Liquid
Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only