missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Borane-tetrahydrofuran complex, 1M solution in THF, Stabilized, AcroSeal™, Thermo Scientific Chemicals
$127.28 - $620.96
Specifications
Chemical Name or Material | Borane-tetrahydrofuran complex |
---|---|
Color | Colorless |
Name Note | 1M solution in tetrahydrofuran, stabilized |
CAS | 14044-65-6,109-99-9 |
Molecular Formula | C4H11BO |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC175081000
|
Thermo Scientific Chemicals
175081000 |
100 mL | AcroSeal™ Glass bottle |
Each for $127.28
|
|
||||
AC175088000
|
Thermo Scientific Chemicals
175088000 |
800 mL | AcroSeal™ Glass bottle |
Each for $620.96
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Borane-tetrahydrofuran complex | |
1M solution in tetrahydrofuran, stabilized | |
C4H11BO | |
0.96 to 1.08M | |
0.8760g/mL | |
borane-tetrahydrofuran complex, tetrahydrofuran borane, bh3.thf, borane tetrahydrofuran complex solution, borane-d3-thf complex solution, borane-tetrahydrofuran, unii-5ear4err1l, oxolane borane, boron; oxolane, borane thf | |
RMCYTHFAWCWRFA-UHFFFAOYSA-N | |
oxolane borane | |
11062302 | |
AcroSeal™ Glass bottle | |
Stabilized | |
Liquid |
Colorless | |
14044-65-6,109-99-9 | |
MFCD00012429 | |
01,199; 02,106; 03,76; 04,124; 05,184; 06,161; 07,89; 12,65; 17,101 | |
15, 1336 | |
Solubility in water: reacts. | |
B.C1CCOC1 | |
85.94 | |
85.94 | |
0.876 | |
−22°C |
RUO – Research Use Only