missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Boron nitride paste, Thermo Scientific Chemicals
Has Resistance Against Molten Metals and Salts, High thermal Conductivity and Dielectric Strength
$96.51 - $1070.34
Chemical Identifiers
CAS | 10043-11-5 |
---|---|
Molecular Formula | BN |
Molecular Weight (g/mol) | 24.82 |
MDL Number | MFCD00011317 |
InChI Key | LNLSXDSWJBUPHM-UHFFFAOYSA-N |
Synonym | boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r |
PubChem CID | 66227 |
ChEBI | CHEBI:50883 |
SMILES | B=N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA4377314
|
Thermo Scientific Chemicals
04377314 |
25 g |
Each for $96.51
|
|
|||||
AA4377322
|
Thermo Scientific Chemicals
04377322 |
100 g |
Each for $267.64
|
|
|||||
AA4377336
|
Thermo Scientific Chemicals
04377336 |
500 g |
Each for $1,070.34
|
|
|||||
Description
Resistance against molten metals and salts, high thermal conductivity and dielectric strength
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10043-11-5 | |
24.82 | |
LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
66227 | |
B=N |
BN | |
MFCD00011317 | |
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
CHEBI:50883 |
Specifications
White | |
Paste | |
BN | |
boron nitride, elbor, borazon, elboron, kubonit, wurzin, denka boron nitride gp, geksanit r, hexanite r | |
B=N | |
24.82 | |
CHEBI:50883 | |
White paste | |
Boron nitride |
10043-11-5 | |
25 g | |
MFCD00011317 | |
LNLSXDSWJBUPHM-UHFFFAOYSA-N | |
azanylidyneborane | |
66227 | |
Odorless | |
1.4 |
Safety and Handling
H319-H335: Causes serious eye irritation. May cause respiratory irritation.
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only