Learn More
Boron tribromide, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 365210250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Lewis acidSpecifications
91.2°C | |
-46.0°C | |
10294-33-4 | |
99+% | |
BBr3 | |
01,66; 02,33; 03,30; 04,42; 05,49; 06,64; 09,61; 10,50; 11,71; 13,43 | |
boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide | |
ILAHWRKJUDSMFH-UHFFFAOYSA-N | |
tribromoborane | |
25134 | |
≥99% | |
2.65 |
Boron tribromide | |
99+% | |
25 g | |
BBr3 | |
MFCD00011312 | |
15, 1349 | |
Solubility in water: reacts. Other solubilities: decomposes in alcohol | |
BrB(Br)Br | |
250.52 | |
250.52 | |
Glass bottle | |
2.6500g/mL |
Chemical Identifiers
10294-33-4 | |
250.52 | |
ILAHWRKJUDSMFH-UHFFFAOYSA-N | |
25134 | |
BrB(Br)Br |
BBr3 | |
MFCD00011312 | |
boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide | |
tribromoborane |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
Fatal if swallowed.
Reacts violently with water.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
EINECSNumber : 233-657-9
Recommended Storage : Poison Room
RUO – Research Use Only