missing translation for 'onlineSavingsMsg'
Learn More

Boron tribromide, 99+%

CAS: 10294-33-4 | BBr3 | 250.52 g/mol

$107.63 - $211.84

Chemical Identifiers

CAS 10294-33-4
Molecular Formula BBr3
Molecular Weight (g/mol) 250.52
MDL Number MFCD00011312
InChI Key ILAHWRKJUDSMFH-UHFFFAOYSA-N
Synonym boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide
PubChem CID 25134
IUPAC Name tribromoborane
SMILES BrB(Br)Br
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC365210250
SDS View Documents
Thermo Scientific Chemicals
365210250
25 g Glass bottle
Each for $107.63
Only null left
 
AC365211000
SDS View Documents
Thermo Scientific Chemicals
365211000
100 g Glass bottle
Each for $211.84
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Lewis acid
Specifications

Chemical Identifiers

10294-33-4
250.52
ILAHWRKJUDSMFH-UHFFFAOYSA-N
25134
BrB(Br)Br
BBr3
MFCD00011312
boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide
tribromoborane

Specifications

91.2°C
10294-33-4
99+%
BBr3
01,66; 02,33; 03,30; 04,42; 05,49; 06,64; 09,61; 10,50; 11,71; 13,43
boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide
ILAHWRKJUDSMFH-UHFFFAOYSA-N
tribromoborane
25134
≥99%
2.65
Boron tribromide, 99+%
-46.0°C
25 g
BBr3
MFCD00011312
15, 1349
Solubility in water: reacts. Other solubilities: decomposes in alcohol
BrB(Br)Br
250.52
250.52
Glass bottle
2.6500g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
Fatal if swallowed.
Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Danger

EINECSNumber : 233-657-9

Recommended Storage : Poison Room

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only