missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Boron tribromide, 99+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 365211000

 View more versions of this product

Catalog No. AC365211000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Lewis acid
Specifications

Specifications

91.2°C
-46.0°C
10294-33-4
99+%
BBr3
01,66; 02,33; 03,30; 04,42; 05,49; 06,64; 09,61; 10,50; 11,71; 13,43
boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide
ILAHWRKJUDSMFH-UHFFFAOYSA-N
tribromoborane
25134
≥99%
2.65
Boron tribromide
99+%
100 g
BBr3
MFCD00011312
15, 1349
Solubility in water: reacts. Other solubilities: decomposes in alcohol
BrB(Br)Br
250.52
250.52
Glass bottle
2.6500g/mL

Chemical Identifiers

10294-33-4
250.52
ILAHWRKJUDSMFH-UHFFFAOYSA-N
25134
BrB(Br)Br
BBr3
MFCD00011312
boron tribromide, borane, tribromo, boron bromide, tribromoboron, hsdb 327, tribromoboran, boron tribrornide, boron tri bromide, boron tri-bromide
tribromoborane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
Fatal if swallowed.
Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Danger

EINECSNumber : 233-657-9

Recommended Storage : Poison Room

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only