Learn More
Boron trifluoride dihydrate, 65% BF3
CAS: 13319-75-0 | BF3H4O2 | 103.84 g/mol
$96.58 - $96.58
Chemical Identifiers
| CAS | 13319-75-0 |
|---|---|
| Molecular Formula | BF3H4O2 |
| Molecular Weight (g/mol) | 103.84 |
| MDL Number | MFCD00149901 |
| InChI Key | MJCYPBSRKLJZTB-UHFFFAOYSA-N |
| Synonym | boron trifluoride dihydrate, boron fluoride dihydrate, borane, trifluoro-, dihydrate, trifluoroborane dihydrate, bf3.2h2o, bf3 dihydrate, bortrifluorid dihydrat, borontrifluoride dihydrate, dsstox_cid_13264, dsstox_rid_79058 |
| PubChem CID | 61591 |
| IUPAC Name | trifluoroborane dihydrate |
| SMILES | O.O.FB(F)F |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC388711000
|
Thermo Scientific Chemicals
388711000 |
100 mL | Glass bottle |
Each for $96.58
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acidChemical Identifiers
| 13319-75-0 | |
| 103.84 | |
| MJCYPBSRKLJZTB-UHFFFAOYSA-N | |
| 61591 | |
| O.O.FB(F)F |
| BF3H4O2 | |
| MFCD00149901 | |
| boron trifluoride dihydrate, boron fluoride dihydrate, borane, trifluoro-, dihydrate, trifluoroborane dihydrate, bf3.2h2o, bf3 dihydrate, bortrifluorid dihydrat, borontrifluoride dihydrate, dsstox_cid_13264, dsstox_rid_79058 | |
| trifluoroborane dihydrate |
Specifications
| 58°C to 60°C (1.5mmHg) | |
| 13319-75-0 | |
| Liquid | |
| 65.0 to 66% | |
| BF3·2H2O | |
| boron trifluoride dihydrate, boron fluoride dihydrate, borane, trifluoro-, dihydrate, trifluoroborane dihydrate, bf3.2h2o, bf3 dihydrate, bortrifluorid dihydrat, borontrifluoride dihydrate, dsstox_cid_13264, dsstox_rid_79058 | |
| MJCYPBSRKLJZTB-UHFFFAOYSA-N | |
| trifluoroborane dihydrate | |
| 61591 | |
| 65% (BF3) | |
| Glass bottle | |
| 1.6420g/mL |
| 6°C | |
| Colorless to Yellow | |
| 100 mL | |
| BF3H4O2 | |
| MFCD00149901 | |
| Solubility in water: miscible | |
| O.O.FB(F)F | |
| 103.84 | |
| 103.85 | |
| Confirmed | |
| 1.642 | |
| Boron trifluoride dihydrate |
Safety and Handling
GHS H Statement
Harmful if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
RUO – Research Use Only