Brady™ Caution: Only Authorized Personnel Allowed... Signs
“Caution: Only Authorized Personnel Allowed...” signs from Brady™ helps to control access to restricted areas to maintain a safe and clean environment.
Supplier:  Brady™ 22921
 View more versions of this product
Catalog No. 19-036097
Description
Description
  • Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
  • Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial and institutional environments.
Specifications
Specifications
ANSI Z535, OSHA 1910.145, ASTM D 1593
1 Ea.
Surface Mount
7 in.
175°F
0.06 in.
25.4 cm
-40°F to 175°F
CAUTION Only Authorized Personnel Allowed To Operate this Equipment
Black on Yellow
Plastic
Non-adhesive
Use in industrial, utility, commercial and institutional environments, common uses include safety signs, utility signs, information signs and equipment warning signs, recommended for both indoor and outdoor use
17.7 cm
-40°F
10 in.
-40°C to 80°C
7 x 10 in.
Authorized Personnel Only Sign