Brady™ Caution Radiation Area Signs
“Caution: Radiation Area” safety signs from Brady™ help to manage and sustain a clean and safe workplace. They are available in a variety of materials and sizes that best fit your application.
Supplier:  Brady™ 45029
 View more versions of this product
Catalog No. 19-038-069
Description
Description
  • Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.
  • Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial and institutional environments.
Specifications
Specifications
ANSI Z535, ASTM D 1593, ASTM D 638, ASTM D 790, ASTM D 646, ASTM D 635
1 Ea.
Non-adhesive
Designed for use in industrial, utility, commercial and institutional environments
193°F
0.1 in.
177.8 mm
-40°F to 193°F
Radiation Area
Black on White
Fiberglass
9000 psi
Premium Fiberglass
10 in.
-40°F
7 in.
-40°C to 90°C
10 x 7 in.
Safety Sign