Brady™ Caution Radiation Area Signs
“Caution: Radiation Area” safety signs from Brady™ help to manage and sustain a clean and safe workplace. They are available in a variety of materials and sizes that best fit your application.
$21.69 - $40.00
Specifications
Quantity | 1 Ea. |
---|---|
Min. Temperature (English) | -40°F |
Displayed Text | Radiation Area |
Product Type | Safety Sign |
Catalog Number | Mfr. No. | Material | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity | |||||
19-038-075
|
Brady™
49017 |
Aluminum |
N/A
|
|
|||||
19-038-069
|
Brady™
45029 |
Fiberglass |
N/A
|
|
|||||
19-038-074
|
Brady™
45166 |
Polyester |
N/A
|
|
|||||
Description
- Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
- Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.
- Durable Non-Reflective Aluminum (B-555) is a rigid aluminum panel with protected graphics designed for use in industrial, utility, commercial and institutional environments.
Specifications
1 Ea. | |
-40°F | |
Radiation Area | |
Safety Sign |