missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Danger: Chlorine Gas Signs
Danger: Chlorine Gas Signs from Brady™ are ideal for use in light duty industrial, utility, commercial and institutional environments.
Supplier: Brady™ 84358
Description
- Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial and institutional environments.
- Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
Specifications
ANSI Z535, OSHA 1910.145 | |
1 Ea. | |
Surface Mount | |
10 in. | |
230°F | |
0.006 to 0.01 in. | |
35.56 cm | |
-40°F to 230°F | |
DANGER Chlorine Gas | |
Red on White |
Polyester | |
Permanent Acrylic | |
Identification of fire alarms, extinguishers and emergency exits | |
25.4 cm | |
-40°F | |
14 in. | |
-40°C to 110°C | |
10 x 14 in. | |
Chemical and Hazardous Materials Sign |