missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Danger: Chlorine Gas Signs
Danger: Chlorine Gas Signs from Brady™ are ideal for use in light duty industrial, utility, commercial and institutional environments.
$14.75 - $37.42
Specifications
Certifications/Compliance | ANSI Z535, OSHA 1910.145 |
---|---|
Quantity | 1 Ea. |
Mounting Type | Surface Mount |
Min. Temperature (English) | -40°F |
Displayed Text | DANGER Chlorine Gas |
Catalog Number | Mfr. No. | Material | Dimensions (L x W) | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Dimensions (L x W) | Price | Quantity | ||||
19-035-701
|
Brady™
84358 |
Polyester | 10 x 14 in. |
Each for $37.42
|
|
||||
19-102-391
|
Brady™
BRD22315 |
Plastic | 7 x 10 in. |
Each for $14.75
|
|
||||
Description
- Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial and institutional environments.
- Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
Specifications
ANSI Z535, OSHA 1910.145 | |
Surface Mount | |
DANGER Chlorine Gas | |
Red on White |
1 Ea. | |
-40°F | |
Chemical and Hazardous Materials Sign |