Brady™ Direction Flow Arrow Tapes
Provide 360° visibility to your indoor and outdoor pipe markers with this durable , pressure-sensitive vinyl (B-946) Direction Flow Arrow Tapes.
Supplier:  Brady™ 91426
 View more versions of this product
Catalog No. 19-017-494
Explore available promotions
Description
Description
  • B-946 Vinyl Arrow Tape is supplied in liner-mounted rolled form - use two bands per pipe marker
  • High gloss, durable, flexible vinyl with a permanent cold temperature pressure sensitive acrylic adhesive
  • Good chemical, oil and water resistance
  • Average outdoor durability 8-10 years
Specifications
Specifications
ASME (ANSI) A13.1 - 2007
White, Red
Tape
Arrow Tape
Pipe marking
60 Gardner Units
1440/Pk.
-40°C to +82°C
30 yd. x 1 in.
27.43 m
25.4 mm
Acrylic, Pressure-sensitive
High Gloss
Vinyl
Non-sterile
Pipe & Valve Marking
8 to 10 Years
-40 to 180°F
0.004 in.
30 yd.
1 in.