Brady™ Direction Flow Arrow Tapes
Provide 360° visibility to your indoor and outdoor pipe markers with this durable , pressure-sensitive vinyl (B-946) Direction Flow Arrow Tapes.
$37.72 - $144.76
Specifications
Adhesive Type Acrylic, Pressure-sensitive
Finish High Gloss
Format Tape
Material Vinyl
Product Type Arrow Tape
View More Specs
Products 10
Catalog Number Mfr. No. Color Width (English) Quantity Price Quantity  
Catalog Number Mfr. No. Color Width (English) Quantity Price Quantity  
17-042-1A
View Documents
Brady™
91424
Black, Yellow 1 in. 1440/Pk.
Each for $40.06
17-042A
View Documents
Brady™
91425
White, Green 1 in. 1440/Pk.
Each for $40.06
17-042B
View Documents
Brady™
91421
White, Green 2 in. 720/Pk.
Each for $80.13
19-017-473
View Documents
Brady™
91288
White, Red 4 in. 270/Pk.
Each for $144.76
19-017-494
View Documents
Brady™
91426
White, Red 1 in. 1440/Pk.
Each for $39.87
19-017-475
View Documents
Brady™
91290
White, Green 4 in. 270/Pk.
Each for $144.76
19-017-472
View Documents
Brady™
91287
Black, Yellow 4 in. 270/Pk.
Each for $144.76
19-017-495
View Documents
Brady™
91427
White, Blue 1 in. 1440/Pk.
Each for $40.06
19-111-883
View Documents
Brady™
91411
Black, Gray 4 in. 270/Pk.
Each for $144.76
19-111-887
View Documents
Brady™
91415
Black, Gray 1 in. 1440/Pk.
Each for $37.72
Description
Description
  • B-946 Vinyl Arrow Tape is supplied in liner-mounted rolled form - use two bands per pipe marker
  • High gloss, durable, flexible vinyl with a permanent cold temperature pressure sensitive acrylic adhesive
  • Good chemical, oil and water resistance
  • Average outdoor durability 8-10 years
Specifications
Specifications
Acrylic, Pressure-sensitive
Tape
Arrow Tape
Pipe marking
8 to 10 Years
-40°C to +82°C
30 yd.
High Gloss
Vinyl
Non-sterile
Pipe & Valve Marking
-40 to 180°F
0.004 in.
27.43 m
SDS
missing translation for 'documents'
missing translation for 'documents'