Brady™ Do Not Open / Do Not Open Valve Tags
“Do Not Open / Do Not Open Valve” accident prevention tags are perfect for marking on-the-spot warnings of dangerous conditions.
Supplier:  Brady™ 86688
 View more versions of this product
Catalog No. 19-040-365
Description
Description
  • Rigid economy polyester and cardstock write-on tags meet OSHA Standard 1910.145 for accident prevention.
  • Durable heavy duty write-on polyester (B-851) tags with brass grommets exceeds OSHA's 50 lb pull force requirement for tagout devices.
  • Withstands moisture, grease and dirt.
  • Paper cardstock (B-853) tags are recommended for indoor use only.
Specifications
Specifications
Accident Prevention Tag
Polyester
4 x 7 in.
Temporary tagging in a non harsh environment
178 mm
4 in.
Black/Red on White
OSHA 1910.145
Header: Danger; Front legend: DO NOT OPEN THIS VALVE; Back legend: DO NOT REMOVE THIS TAG
7 in.
0.25 mm
102 mm