Brady™ Fuse Lockout Device
These Fuse Lockout Devices from Brady™ have a unique locking mechanism that prevents the device from being accidentally removed from the fuse block.
Supplier: Brady™ 65750
Description
- Equipping your employees with the proper lockout tools and warning devices can save lives, reduce lost employee time, and cut insurance costs
- Brady is a world leader in lockout/tagout products and services, offering industry expertise and a broad range of durable, easy-to-use lockout devices
Specifications
Fuse Lockout Device | |
ABS | |
Fuse size 0.4 in. | |
Small |
Red | |
OSHA 1910.147 | |
6 plastic nonconductive lockouts for use with cable ties and 0.4 in. fuses |