Brady™ General Traffic Signs
Brady traffic signs provide direction or information to sustain a safe work environment.
Supplier:  Brady™ 94145
 View more versions of this product
Catalog No. 18-999-4322
Explore available promotions
Description
Description
  • Durable Reflective Aluminum (B-959) is constructed of reflective sheeting applied to an aluminum sign panel and is ideal for traffic control applications.
  • Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.
Specifications
Specifications
MUTCD R1-1
1 Ea.
Reflective Aluminum
36 in.
-40°F
36 in.
-40°C to 80°C
36 x 36 in.
Traffic Sign
Aluminum
Non-adhesive
Traffic control
175°F
0.04 to 0.09 in.
91.44 cm
-40°F to 175°F
Stop
Red on White