Brady™ Globally Harmonized System (GHS) Acute Toxic Picto Labels
For acute toxicity, skin and eye irritation, skin sensitizers
Supplier:  Brady™ 118857
 View more versions of this product
Catalog No. 19-140-519
Explore available promotions
Description
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
  • Legend: Acute Toxic
  • Made of B-7569 Viny or B-235 Paper
  • Available in three sizes
Specifications
Specifications
GHS, OSHA 29 CFR 1910.1200
Black, Red, White
Pictogram Label
For acute toxicity, skin and eye irritation, skin sensitizers
0.625 x 0.625 in.
-4 to 167°F
0.6 in.
15.20 mm
Acrylic, Pressure-sensitive
Vinyl
Acute Toxic
39/Pk.
1.58 x 1.58 cm (W x H)
-20°C to +75°C
0.62 in.