missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Globally Harmonized System (GHS) Acute Toxic Picto Labels
For acute toxicity, skin and eye irritation, skin sensitizers
$10.73 - $14.07
Specifications
Adhesive Type | Acrylic, Pressure-sensitive |
---|---|
Color | Black, Red, White |
Display Text | Acute Toxic |
For Use With (Application) | For acute toxicity, skin and eye irritation, skin sensitizers |
Catalog Number | Mfr. No. | Quantity | Material | Size (Metric) | Size (English) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Material | Size (Metric) | Size (English) | Price | Quantity | |||
19-140-519
|
Brady™
118857 |
39/Pk. | Vinyl | 1.58 x 1.58 cm (W x H) | 0.625 x 0.625 in. |
Pack of 39 Each for $14.07
|
|
|||
19-140-528
|
Brady™
118911 |
39/Pk. | Paper | 1.58 x 1.58 cm (W x H) | 0.625 x 0.625 in. |
Pack of 39 Each for $11.09
|
|
|||
19-140-537
|
Brady™
118858 |
16/Pk. | Vinyl | 2.54 x 2.54 cm | 1 x 1 in. |
Pack of 16 Each for $12.91
|
|
|||
19-140-546
|
Brady™
118912 |
16/Pk. | Paper | 2.54 x 2.54 cm | 1 x 1 in. |
Pack of 16 Each for $10.73
|
|
|||
19-140-706
|
Brady™
118859 |
4/Card | Vinyl (B-7569) | 5.08 x 5.08 cm | 2 x 2 in. |
Pack of 4 Each for $12.05
|
|
|||
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
- Legend: Acute Toxic
- Made of B-7569 Viny or B-235 Paper
- Available in three sizes
Specifications
Acrylic, Pressure-sensitive | |
Black, Red, White | |
Acute Toxic | |
For acute toxicity, skin and eye irritation, skin sensitizers |