Brady™ Globally Harmonized System (GHS) Acute Toxic Picto Labels
For acute toxicity, skin and eye irritation, skin sensitizers
Supplier:  Brady™ 118858
 View more versions of this product
Catalog No. 19-140-537
Explore available promotions
Description
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
  • Legend: Acute Toxic
  • Made of B-7569 Viny or B-235 Paper
  • Available in three sizes
Specifications
Specifications
GHS, OSHA 29 CFR 1910.1200
Black, Red, White
Pictogram Label
For acute toxicity, skin and eye irritation, skin sensitizers
1 x 1 in.
-4 to 167°F
0.004 in.
1 in.
Acrylic, Pressure-sensitive
Vinyl
Acute Toxic
16/Pk.
2.54 x 2.54 cm
-20°C to +75°C
1 in.
2.5 cm