missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Globally Harmonized System (GHS) Health Danger Picto Labels
Indicates carcinogens, respiratory sensitizers, reproductive toxicity, target organ toxicity, germ cell mutagens
Supplier: Brady™ 118852
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations– universal approach to classifying and communicating chemical hazards
- Legend: Health Danger
- Made of B-7569 viny or B-235 paper
- Available in three sizes
Specifications
GHS, OSHA 29 CFR 1910.1200 | |
Black, Red, White | |
Pictogram Label | |
For carcinogens, respiratory sensitizers, reproductive toxicity, target organ toxicity, germ cell mutagens | |
-4 to 167°F | |
0.004 in. | |
1 in. |
Acrylic, Pressure-sensitive | |
Vinyl | |
Health Danger | |
16/Pk. | |
-20°C to +75°C | |
1 in. | |
2.5 cm |