missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Globally Harmonized System (GHS) Health Danger Picto Labels
Indicates carcinogens, respiratory sensitizers, reproductive toxicity, target organ toxicity, germ cell mutagens
$11.27 - $14.61
Specifications
Certifications/Compliance | GHS, OSHA 29 CFR 1910.1200 |
---|---|
Color | Black, Red, White |
For Use With (Application) | For carcinogens, respiratory sensitizers, reproductive toxicity, target organ toxicity, germ cell mutagens |
Catalog Number | Mfr. No. | Quantity | Material | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Material | Price | Quantity | ||||
19-140-518
|
Brady™
118851 |
39/Pk. | Vinyl |
Each for $14.61
|
|
||||
19-140-527
|
Brady™
118905 |
39/Pk. | Paper |
Each for $11.64
|
|
||||
19-140-536
|
Brady™
118852 |
16/Pk. | Vinyl |
Each for $14.61
|
|
||||
19-140-545
|
Brady™
118906 |
16/Pk. | Paper |
Each for $11.27
|
|
||||
19-140-705
|
Brady™
118853 |
4/Pk. | Vinyl (B-7569) |
Pack of 4 Each for $12.59
|
|
||||
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations– universal approach to classifying and communicating chemical hazards
- Legend: Health Danger
- Made of B-7569 viny or B-235 paper
- Available in three sizes
Specifications
GHS, OSHA 29 CFR 1910.1200 | |
Black, Red, White | |
For carcinogens, respiratory sensitizers, reproductive toxicity, target organ toxicity, germ cell mutagens |