missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Globally Harmonized System (GHS) Severe Toxic Picto Labels
For severe toxicity
Supplier: Brady™ 118899
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
- Legend: Severe Toxic
- Made of B-7569 Viny or B-235 Paper
- Available in three sizes
Specifications
GHS, OSHA 29 CFR 1910.1200 | |
Black, Red, White | |
Pictogram Label | |
For severe toxicity | |
-40 to 150°F | |
0.0038 in. | |
0.62 in. |
Acrylic, Pressure-sensitive | |
Paper (B-235) | |
Severe Toxic | |
39/Pk. | |
-40°C to +65°C | |
0.6 in. | |
15.20 mm |