missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Globally Harmonized System (GHS) Severe Toxic Picto Labels
For severe toxicity
$11.27 - $14.61
Specifications
Certifications/Compliance | GHS, OSHA 29 CFR 1910.1200 |
---|---|
Color | Black, Red, White |
For Use With (Application) | For severe toxicity |
Catalog Number | Mfr. No. | Material | Adhesive Type | Width (English) | Height (English) | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Adhesive Type | Width (English) | Height (English) | Price | Quantity | ||
19-140-517
|
Brady™
118845 |
Vinyl (B-7569) | Acrylic, Pressure-sensitive | 0.6 in. | 0.62 in. |
Each for $14.61
|
|
||
19-140-526
|
Brady™
118899 |
Paper (B-235) | Acrylic, Pressure-sensitive | 0.6 in. | 0.62 in. |
Each for $11.27
|
|
||
19-140-535
|
Brady™
118846 |
Vinyl (B-7569) | Acrylic, Pressure-sensitive | 1 in. | 1 in. |
Each for $13.41
|
|
||
19-140-544
|
Brady™
118900 |
Paper (B-235) | Acrylic, Pressure-sensitive | 1 in. | 1 in. |
Each for $11.27
|
|
||
19140704
|
Brady™
118847 |
Vinyl (B-7569) | Pressure-sensitive Acrylic | 2 in. |
Pack of 4 Each for $13.83
|
|
|||
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
- Legend: Severe Toxic
- Made of B-7569 Viny or B-235 Paper
- Available in three sizes
Specifications
GHS, OSHA 29 CFR 1910.1200 | |
Black, Red, White | |
For severe toxicity |