missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Globally Harmonized System (GHS) Severe Toxic Picto Labels
For severe toxicity
Supplier: Brady™ 118900
Description
Pre-printed die cut labels identify hazardous products for chemical risks, health risks and environmental risks in accordance to the United Nations' universal approach to classifying and communicating chemical hazards
- Legend: Severe Toxic
- Made of B-7569 Viny or B-235 Paper
- Available in three sizes
Specifications
| GHS, OSHA 29 CFR 1910.1200 | |
| Black, Red, White | |
| Label | |
| Pictogram Label | |
| Severe Toxic | |
| 16/Pk. | |
| -40°C to +65°C | |
| 1 x 1 in. | |
| 1 in. |
| Acrylic, Pressure-sensitive | |
| Semi-gloss | |
| Paper (B-235) | |
| Non-sterile | |
| For severe toxicity | |
| -40°F to 150°F | |
| 0.0038 in. | |
| 1 in. | |
| 2.5 cm |